CH6371390246 Chem Impex International, Inc.

3-Bromo-4-nitrobenzoic acid methyl ester

  • MDL number MFCD11045599
  • Cas Number : 126759-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-77
Synonym Methyl 3-bromo-4-nitrobenzoate
Purity 98
Storage temp. Store at RT
Physical Form White to yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-bromo-4-nitrobenzoic-acid-methyl-ester/19303?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-bromo-4-nitrobenzoic-acid-methyl-ester/19303?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart