CH6447916305 Chem Impex International, Inc.

5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine

  • MDL number MFCD00010059
  • Cas Number : 68892-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5'-DMT-NiBu-dG
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-i-n-i-sup-2-sup-isobutyryl-2-deoxyguanosine/117?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-i-n-i-sup-2-sup-isobutyryl-2-deoxyguanosine/117?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart