CH6447916319 Chem Impex International, Inc.

5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine-3'-lcaa-CPG

  • MDL number MFCD01310317
  • Molecular Weight : 638.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at RT
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-o-4-4-dimethoxytrityl-n-sup-2-sup-isobutyryl-2-deoxyguanosine-3-lcaa-cpg/332?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-o-4-4-dimethoxytrityl-n-sup-2-sup-isobutyryl-2-deoxyguanosine-3-lcaa-cpg/332?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart