CH6447916320 Chem Impex International, Inc.

5'-O-(4,4'-Dimethoxytrityl)-N4-benzoyl-2'-deoxycytidine-3'-lcaa-CPG

  • MDL number MFCD01310318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DMT-dC-NBz-lcaa-CPG, 500A, 20-40 �
Storage temp. Store at RT
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-o-4-4-dimethoxytrityl-n-sup-4-sup-benzoyl-2-deoxycytidine-3-lcaa-cpg/333?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-o-4-4-dimethoxytrityl-n-sup-4-sup-benzoyl-2-deoxycytidine-3-lcaa-cpg/333?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart