CH6447916325 Chem Impex International, Inc.

L-Tyrosine disodium salt dihydrate, Non-animal/Non-human Origin

  • MDL number MFCD03840558
  • Cas Number : 122666-87-9
  • Molecular Weight : 261.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at RT
Physical Form Yellowish to tan powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-tyrosine-disodium-salt-dihydrate/364?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-tyrosine-disodium-salt-dihydrate/364?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart