CH6447916329 Chem Impex International, Inc.

b-Nicotinamide adenine dinucleotide phosphate disodium salt dihdydrate

  • MDL number MFCD22495363
  • Cas Number : 24292-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym NADP 2Na 2H2O
Purity 0.98
Storage temp. Store at = - 15
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-nicotinamide-adenine-dinucleotide-phosphate-disodium-salt-dihdydrate/416?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-nicotinamide-adenine-dinucleotide-phosphate-disodium-salt-dihdydrate/416?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart