CH6447916454 Chem Impex International, Inc.

O-Benzyl-D-tyrosine

  • MDL number MFCD00077104
  • Cas Number : 65733-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Tyr(Bzl)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-d-tyrosine/2251?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-d-tyrosine/2251?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart