CH6447916554 Chem Impex International, Inc.

N-(2-Chlorotrityl resin)-S-4-methoxytrityl-L-cysteine benzotriazolyl ester

  • MDL number MFCD01074995

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-i-s-i-4-methoxytrityl-l-cysteine-benzotriazolyl-ester/2926?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-i-s-i-4-methoxytrityl-l-cysteine-benzotriazolyl-ester/2926?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart