CH6447916555 Chem Impex International, Inc.

N-(2-Chlorotrityl resin)-glycine N-hydroxysuccinimide ester

  • MDL number MFCD01074996

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-glycine-i-n-i-hydroxysuccinimide-ester/2927?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-glycine-i-n-i-hydroxysuccinimide-ester/2927?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart