CH6447916556 Chem Impex International, Inc.

2-Chlorotrityl resin-L-Leu-OBt

  • MDL number MFCD01074997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chlorotrityl resin-L-Leu-OBt
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-l-leucine-benzotriazolyl-ester/2928?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-l-leucine-benzotriazolyl-ester/2928?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart