CH6447916557 Chem Impex International, Inc.

Na-(2-Chlorotrityl resin)-Ne-Boc-L-lysine benzotriazolyl ester

  • MDL number MFCD01074998

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chlorotrityl resin-L-Lys(Boc)-OBt
Storage temp. Store at 0-8
Physical Form Yellow
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-2-chlorotrityl-resin-i-n-i-sup-epsilon-sup-boc-l-lysine-benzotriazolyl-ester/2929?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-2-chlorotrityl-resin-i-n-i-sup-epsilon-sup-boc-l-lysine-benzotriazolyl-ester/2929?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart