CH6447916560 Chem Impex International, Inc.

N-(2-Chlorotrityl resin)-O-trityl-L-serine benzotriazolyl ester

  • MDL number MFCD01075001

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chlorotrityl resin-L-Ser(Trt)-OBt
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-i-o-i-trityl-l-serine-benzotriazolyl-ester/2932?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-i-o-i-trityl-l-serine-benzotriazolyl-ester/2932?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart