CH6447916561 Chem Impex International, Inc.

N-(2-Chlorotrityl resin)-L-valine benzotriazolyl ester

  • MDL number MFCD01075002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chlorotrityl resin-L-Val-OBt
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-l-valine-benzotriazolyl-ester/2933?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-2-chlorotrityl-resin-l-valine-benzotriazolyl-ester/2933?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart