CH6447916562 Chem Impex International, Inc.

2-Chlorotrityl resin-4-aminomethylbenzoic acid-OBt

  • MDL number MFCD01075003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(2-Chlorotrityl resin)-4-aminomethylbenzoic acid benzotriazolyl ester
Storage temp. Store at RT
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chlorotrityl-resin-4-aminomethylbenzoic-acid-obt/2934?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chlorotrityl-resin-4-aminomethylbenzoic-acid-obt/2934?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart