CH6447916575 Chem Impex International, Inc.

1,4-Diaminobutane trityl resin

  • MDL number MFCD03414208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Physical Form Yellow to brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-4-diaminobutane-trityl-resin/2947?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-4-diaminobutane-trityl-resin/2947?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart