CH6447916583 Chem Impex International, Inc.

Acetyl-L-tyrosine methyl amide

  • MDL number MFCD00237518
  • Cas Number : 6367-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-tyrosine-methyl-amide/3027?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-tyrosine-methyl-amide/3027?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart