CH6447916584 Chem Impex International, Inc.

L-Aspartic acid a-amide hydrochloride

  • MDL number MFCD06795769
  • Cas Number : 200260-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-180
Synonym L-Asp-NH2 HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-alpha-amide-hydrochloride/3041?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-alpha-amide-hydrochloride/3041?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart