CH6447916587 Chem Impex International, Inc.

Val-Gly-NH2 HCl

  • MDL number MFCD00155521
  • Cas Number : 36920-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/val-gly-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hcl/3128?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/val-gly-nh-sub-2-sub-font-size-2-face-symbol-middot-font-hcl/3128?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart