CH6447916614 Chem Impex International, Inc.

Fmoc-3-cyclohexyl-L-alanine 4-alkoxybenzyl alcohol resin

  • MDL number MFCD00238744

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Cha-Wang resin
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-cyclohexyl-l-alanine-4-alkoxybenzyl-alcohol-resin/3398?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-cyclohexyl-l-alanine-4-alkoxybenzyl-alcohol-resin/3398?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart