CH6447916616 Chem Impex International, Inc.

Nalpha-Fmoc-Nbeta-Boc-L-2,3-diaminopropionic acid 4-alkoxybenzyl alcohol resin

  • MDL number MFCD01310964

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Physical Form Pale to yellow beads
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-beta-sup-boc-l-2-3-diaminopropionic-acid-4-alkoxybenzyl-alcohol-resin/3400?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-beta-sup-boc-l-2-3-diaminopropionic-acid-4-alkoxybenzyl-alcohol-resin/3400?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart