CH6447916617 Chem Impex International, Inc.

Fmoc-Ala(2-naphthyl)-Wang resin

  • MDL number MFCD01310943

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-ala-2-naphthyl-wang-resin/3401?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-ala-2-naphthyl-wang-resin/3401?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart