CH6447916619 Chem Impex International, Inc.

Fmoc-4-fluoro-D-phenylalanine 4-alkoxybenzyl alcohol resin

  • MDL number MFCD01310951

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-fluoro-d-phenylalanine-4-alkoxybenzyl-alcohol-resin/3403?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-fluoro-d-phenylalanine-4-alkoxybenzyl-alcohol-resin/3403?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart