CH6447916620 Chem Impex International, Inc.

Fmoc-L-pipecolic acid 4-alkoxybenzyl alcohol resin

  • MDL number MFCD01310965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Pip-Wang resin
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-pipecolic-acid-4-alkoxybenzyl-alcohol-resin/3404?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-pipecolic-acid-4-alkoxybenzyl-alcohol-resin/3404?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart