CH6447916625 Chem Impex International, Inc.

Boc-Ala-Phe-OH

  • MDL number MFCD00235762

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-ala-phe-oh/3416?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-ala-phe-oh/3416?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart