CH6447916626 Chem Impex International, Inc.

Boc-Trp-Phe-OMe

  • MDL number MFCD00235772
  • Cas Number : 72156-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-trp-phe-ome/3424?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-trp-phe-ome/3424?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart