CH6447916633 Chem Impex International, Inc.

Fmoc-L-phenylglycine 4-alkoxybenzyl alcohol resin

  • MDL number MFCD01310953

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-phenylglycine-4-alkoxybenzyl-alcohol-resin/3483?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-phenylglycine-4-alkoxybenzyl-alcohol-resin/3483?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart