CH6447916637 Chem Impex International, Inc.

Na-Boc-Na-methyl-Ne-Z-L-lysine dicyclohexylammonium salt

  • MDL number MFCD00236866

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-N-Me-L-Lys(Z)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-alpha-sup-methyl-i-n-i-sup-epsilon-sup-z-l-lysine-dicyclohexylammonium-salt/3646?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-alpha-sup-methyl-i-n-i-sup-epsilon-sup-z-l-lysine-dicyclohexylammonium-salt/3646?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart