CH6447916638 Chem Impex International, Inc.

Fmoc-Pro-Gly-OH

  • MDL number MFCD00237671
  • Cas Number : 250695-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-163
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-pro-gly-oh/19824?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-pro-gly-oh/19824?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart