CH6447916639 Chem Impex International, Inc.

DL-alpha-Methyltryptophan methyl ester hydrochloride

  • MDL number MFCD00152138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-144 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-alpha-methyltryptophan-methyl-ester-hydrochloride/3804?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-alpha-methyltryptophan-methyl-ester-hydrochloride/3804?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart