CH6447916642 Chem Impex International, Inc.

L-Glutamine alpha-4-nitrobenzyl ester hydrobromide

  • MDL number MFCD00136774
  • Cas Number : 14349-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-169 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamine-alpha-4-nitrobenzyl-ester-hydrobromide/3961?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamine-alpha-4-nitrobenzyl-ester-hydrobromide/3961?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart