CH6447916646 Chem Impex International, Inc.

H-Phe-Ser-OH

  • MDL number MFCD00083787
  • Cas Number : 16053-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/h-phe-ser-oh/19822?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-phe-ser-oh/19822?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart