CH6447916649 Chem Impex International, Inc.

b-Chloro-D-alanine

  • MDL number MFCD01632068
  • Cas Number : 51887-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-157
Synonym D-beta-Chloro-Ala-OH
Purity 99
Storage temp. Store at = -20
Physical Form White fine needles
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-chloro-d-alanine/4083?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-chloro-d-alanine/4083?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart