CH6447916650 Chem Impex International, Inc.

N-Methyl-L-aspartic acid hydrochloride

  • MDL number MFCD06795788
  • Cas Number : 4226-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-L-Asp-OH HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-aspartic-acid-hydrochloride/4168?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-aspartic-acid-hydrochloride/4168?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart