CH6447916652 Chem Impex International, Inc.

N-Methyl-L-glutamic acid

  • MDL number MFCD00065920
  • Cas Number : 6753-62-4
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-glutamic-acid/4176?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-glutamic-acid/4176?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart