CH6447916653 Chem Impex International, Inc.

N-Methyl-L-tyrosine

  • MDL number MFCD00238322
  • Cas Number : 537-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-L-Tyr-OH
Purity 95
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-tyrosine/4190?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-tyrosine/4190?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart