CH6447916654 Chem Impex International, Inc.

Na-Z-Ng-trityl-D-asparagine

  • MDL number MFCD00237334
  • Cas Number : 200259-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-191 �C
Synonym Z-D-Asn(Trt)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-gamma-sup-trityl-d-asparagine/4235?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-gamma-sup-trityl-d-asparagine/4235?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart