CH6447916655 Chem Impex International, Inc.

Z-Glu-Gly-OH

  • MDL number MFCD00191093
  • Cas Number : 1634-89-5
  • Molecular Weight : 338.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-149 °C
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-glu-gly-oh/19830?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-glu-gly-oh/19830?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart