CH6447916664 Chem Impex International, Inc.

Acetyl-S-farnesyl-L-cysteine

  • MDL number MFCD00871540
  • Cas Number : 135304-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Cys(Farnesyl)-OH
Purity 99
Storage temp. Store at = -4
Physical Form Yellow-red oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-i-s-i-farnesyl-l-cysteine/4460?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-i-s-i-farnesyl-l-cysteine/4460?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart