CH6447916682 Chem Impex International, Inc.

Decanoyl-Arg-Val-Lys-Arg-CMK�

  • MDL number MFCD18910843
  • Cas Number : 150113-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Furin Inhibitor I Trifluoroacetate
Purity 90
Storage temp. Store at = -4
Physical Form White powder to lyophilisate
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/decanoyl-arg-val-lys-arg-cmk-b-font-size-2-face-symbol-middot-font-b-tfa/4896?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/decanoyl-arg-val-lys-arg-cmk-b-font-size-2-face-symbol-middot-font-b-tfa/4896?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart