CH6447916685 Chem Impex International, Inc.

Ng-(2-Acetamido-2-deoxy-b-D-gluco-pyranosyl)-L-asparagine

  • MDL number MFCD02259695
  • Cas Number : 2776-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Asn(GlcNAc-beta-D)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-2-acetamido-2-deoxy-ß-d-gluco-pyranosyl-l-asparagine/4947?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-2-acetamido-2-deoxy-ß-d-gluco-pyranosyl-l-asparagine/4947?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart