CH6447916690 Chem Impex International, Inc.

N-Me-DL-phenylglycine

  • MDL number MFCD01318769
  • Cas Number : 74641-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-DL-Phg-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-me-dl-phenylglycine/5103?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-me-dl-phenylglycine/5103?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart