CH6447916691 Chem Impex International, Inc.

Succinyl-L-proline

  • MDL number MFCD00057237
  • Cas Number : 63250-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-103 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/succinyl-l-proline/5170?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/succinyl-l-proline/5170?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart