CH6447916698 Chem Impex International, Inc.

1,2-Dipalmitoyl-sn-glycero-3-phosphatidic acid, sodium salt

  • MDL number MFCD00153887
  • Cas Number : 71065-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta,gamma-Dipalmitoyl-alpha-phosphatidic acid Na
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-dipalmitoyl-i-sn-i-glycero-3-phosphatidic-acid-sodium-salt/5306?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-dipalmitoyl-i-sn-i-glycero-3-phosphatidic-acid-sodium-salt/5306?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart