CH6447916699 Chem Impex International, Inc.

Cyclo(-D-Ala-L-Pro)

  • MDL number MFCD00236940
  • Cas Number : 36238-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at = -4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-d-ala-l-pro/5334?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-d-ala-l-pro/5334?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart