CH6447916704 Chem Impex International, Inc.

Boc-b-cyclopropyl-D-alanine�A

  • MDL number MFCD02259470
  • Cas Number : 89483-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-161
Synonym Boc-D-Ala(beta-cyclopropyl)-OH dicyclohexylammonium salt
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-beta-cyclopropyl-d-alanine-font-size-2-face-symbol-middot-font-dcha/5457?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-beta-cyclopropyl-d-alanine-font-size-2-face-symbol-middot-font-dcha/5457?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart