CH6447916712 Chem Impex International, Inc.

Z-L-glutamic acid gamma-cyclohexyl ester dicyclohexylammonium salt

  • MDL number MFCD19707054
  • Cas Number : 111479-43-7
  • Molecular Weight : 544.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 130 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-cyclohexyl-ester-dicyclohexylammonium-salt/5743?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-cyclohexyl-ester-dicyclohexylammonium-salt/5743?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart