CH6447916713 Chem Impex International, Inc.

Nalpha-Z-Nepsilon-4-toluenesulfonyl-L-lysine dicyclohexylammonium salt

  • MDL number MFCD09265179
  • Cas Number : 2362-45-0
  • Molecular Weight : 615.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-i-n-i-sup-e-sup-4-toluenesulfonyl-l-lysine-dicyclohexylammonium-salt/5748?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-i-n-i-sup-e-sup-4-toluenesulfonyl-l-lysine-dicyclohexylammonium-salt/5748?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart