CH6447916714 Chem Impex International, Inc.

D-Ala-D-Leu-OH�

  • MDL number MFCD26793858
  • Cas Number : 1820579-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-147
Synonym D-Alanyl-D-leucine�
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-ala-d-leu-oh-b-font-size-2-face-symbol-middot-font-b-tfa/5794?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-ala-d-leu-oh-b-font-size-2-face-symbol-middot-font-b-tfa/5794?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart