CH6447916723 Chem Impex International, Inc.

Na-Boc-N-p-benzyloxymethyl-L-histidine N-hydroxysuccinimide ester

  • MDL number MFCD00236853
  • Cas Number : 129672-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-83
Synonym Boc-L-His(p-Bom)-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-pi-benzyloxymethyl-l-histidine-i-n-i-hydroxysuccinimide-ester/6064?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-pi-benzyloxymethyl-l-histidine-i-n-i-hydroxysuccinimide-ester/6064?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart