CH6447916728 Chem Impex International, Inc.

DL-Phenylalanine 4-nitroanilide�rochloride salt

  • MDL number MFCD00508152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Phe-pNA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-phenylalanine-4-nitroanilide/6207?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-phenylalanine-4-nitroanilide/6207?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart